Få mere at vide
Cloxacillin Sodium Salt, TRC
Artikelnummer. 30012539
Skift visning
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012539 10 g
30003806 5 g
17924765 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012539 Leverandør Toronto Research Chemicals Leverandørnr. TRCC58746010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 642-78-4
Molekylær formel C19 H17 Cl N3 O5 S . Na
Molekylvægt (g/mol) 457.86
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[[3-(2-chlorophenyl)-5-methyl-4-isoxazolyl]carbonyl]amino]-3,3-dimethyl-7-oxo-, sodium salt (1:1), (2S,5R,6R)-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[3-(o-chlorophenyl)-5-methyl-4-isoxazolecarboxamido]-3,3-dimethyl-7-oxo-, monosodium salt (8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[[3-(2-chlorophenyl)-5-methyl-4-isoxazolyl]carbonyl]amino]-3,3-dimethyl-7-oxo-, monosodium salt, (2S,5R,6R)- (9CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[[3-(2-chlorophenyl)-5-methyl-4-isoxazolyl]carbonyl]amino]-3,3-dimethyl-7-oxo-, monosodium salt, [2S-(2α,5α,6β)]-, 3-(o-Chlorophenyl)-5-methyl-4-isoxazolylpenicillin sodium salt, Ankerbin, Austrastaph, BRL 1621 sodium salt, Cloxacillin sodium, Cloxacillin sodium salt, Cloxapen, Ekvacillin, Gelstaph, Monosodium cloxacillin, Orbenin, Orbenin sodium, Prevencilina P, Prostaphilin A, Prostaphlin A, Sodium 3-(O-chlorophenyl)-5-methyl-4-isoxazolylpenicillin, Sodium cloxacillin, Sodium orbenin, Sodium syntarpen, Staphybiotic, Syntarpen sodium salt, Tegopen, BRL-1621
IUPAC navn sodium;(2S,5R,6R)-6-[[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMIL [Na+].Cc1onc(c2ccccc2Cl)c1C(=O)N[C@H]3[C@H]4SC(C)(C)[C@@H](N4C3=O)C(=O)[O-]
Tekniske data
Kemisk navn eller materiale Cloxacillin Sodium
Mængde 10 g
InChI formel InChI=1S/C19H18ClN3O5S.Na/c1-8-11(12(22-28-8)9-6-4-5-7-10(9)20)15(24)21-13-16(25)23-14(18(26)27)19(2,3)29-17(13)23;/h4-7,13-14,17H,1-3H3,(H,21,24)(H,26,27);/q;+1/p-1/t13-,14+,17-;/m1./s1
Formel vægt 457.0475