missing translation for 'onlineSavingsMsg'
Få mere at vide

Clothianidin-d3 (N-methyl-d3), TRC

Artikelnummer. 30366305
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.5 mg
1 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366305 5 mg
30362407 1 mg
30358039 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366305 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5885015MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1262776-24-8
Molekylær formel C6 2H3 H5 Cl N5 O2 S
Molekylvægt (g/mol) 252.7
Synonym 1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-2-nitro-3-(trideuteriomethyl)guanidine
IUPAC navn 1-[(2-chloro-1,3-thiazol-5-yl)methyl]-2-nitro-3-(trideuteriomethyl)guanidine
SMIL [2H]C([2H])([2H])N\C(=N/[N+](=O)[O-])\NCc1cnc(Cl)s1

Specifications

Kemisk navn eller materiale Clothianidin D3 (N'-methyl D3)
Mængde 5 mg
InChI formel InChI=1S/C6H8ClN5O2S/c1-8-6(11-12(13)14)10-3-4-2-9-5(7)15-4/h2H,3H2,1H3,(H2,8,10,11)/i1D3
Formel vægt 252.0276
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.