Få mere at vide
Clorprenaline Hydrochloride, TRC
Artikelnummer. 30350330
Skift visning
Mængde:
500 mg
1 g
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30350330 2.5 g 2.5g
30363416 500 mg -
30350614 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30350330 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4279552.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6933-90-0
Molekylær formel C11 H16 Cl N O . Cl H
Molekylvægt (g/mol) 250.16
Synonym Benzenemethanol, 2-chloro-α-[[(1-methylethyl)amino]methyl]-, hydrochloride (9CI), Benzyl alcohol, o-chloro-α-[(isopropylamino)methyl]-, hydrochloride (6CI,7CI,8CI), Chlorprenaline hydrochloride, Clorprenaline hydrochloride, Isoprophenamine hydrochloride, N-[2-(2-Chlorophenyl)-2-hydroxyethyl]propan-2-aminium chloride, NSC 334693
IUPAC navn 1-(2-chlorophenyl)-2-(propan-2-ylamino)ethanol;hydrochloride
SMIL Cl.CC(C)NCC(O)c1ccccc1Cl
Tekniske data
Kemisk navn eller materiale Clorprenaline Hydrochloride
Mængde 2.5 g
InChI formel InChI=1S/C11H16ClNO.ClH/c1-8(2)13-7-11(14)9-5-3-4-6-10(9)12;/h3-6,8,11,13-14H,7H2,1-2H3;1H
Formel vægt 249.0687