Få mere at vide
Clorprenaline Hydrochloride, Mikromol™
Beskrivelse
Clorprenaline Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Clorprenaline Hydrochloride |
| CAS | 6933-90-0 |
| Analyt- eller komponentnavne | Clorprenaline Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C11 H16 Cl N O . Cl H |
| Certificeringer/compliance | ISO/IEC 17025 |
| Synonym | Chlorprenaline hydrochloride, Clorprenaline hydrochloride, Isoprophenamine hydrochloride, N-[2-(2-Chlorophenyl)-2-hydroxyethyl]propan-2-aminium chloride, NSC 334693, Benzenemethanol, 2-chloro-alpha-[[(1-methylethyl)amino]methyl]-, hydrochloride (1:1), Benzenemethanol, 2-chloro-alpha-[[(1-methylethyl)amino]methyl]-, hydrochloride (9CI), Benzyl alcohol, o-chloro-alpha-[(isopropylamino)methyl]-, hydrochloride (6CI,7CI,8CI) |
| InChI formel | InChI=1S/C11H16ClNO.ClH/c1-8(2)13-7-11(14)9-5-3-4-6-10(9)12;/h3-6,8,11,13-14H,7H2,1-2H3;1H |
| SMIL | Cl.CC(C)NCC(O)c1ccccc1Cl |
| IUPAC navn | 1-(2-chlorophenyl)-2-(propan-2-ylamino)ethanol;hydrochloride |
| Vis mere |