missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Clorprenaline hydrochloride, MedChemExpress
Beskrivelse
Clorprenaline hydrochloride is a β2-adrenergic receptor agonist that is implicated in bronchial expansion. Clorprenaline has the potential for asthma research.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Clorprenaline hydrochloride |
| CAS | 6933-90-0 |
| Farve | White |
| Renhedsgrad noter | Research |
| Molekylær formel | C11H17Cl2NO |
| Mængde | 50 mg |
| Opløselighedsinformation | DMSO : 100 mg/mL (399.74 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (399.74 mM) |
| SMIL | OC(CNC(C)C)C1=CC=CC=C1Cl.[H]Cl |
| Molekylvægt (g/mol) | 250.16 |
| Formel vægt | 250.16 |
| Vis mere |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?