Få mere at vide
Clorprenaline Hydrochloride, LoGiCal
Artikelnummer. 30458356
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30458356 10 mg
1 options
Artikelnummer. 30458356 Leverandør LoGiCal Leverandørnr. LGCFOR0716.02
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Clorprenaline Hydrochloride
CAS 6933-90-0
Analyt- eller komponentnavne Clorprenaline Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C11 H16 Cl N O . Cl H
Certificeringer/compliance ISO 9001
Synonym Chlorprenaline hydrochloride, Clorprenaline hydrochloride, Isoprophenamine hydrochloride, N-[2-(2-Chlorophenyl)-2-hydroxyethyl]propan-2-aminium chloride, NSC 334693, Benzenemethanol, 2-chloro-alpha-[[(1-methylethyl)amino]methyl]-, hydrochloride (1:1), Benzenemethanol, 2-chloro-alpha-[[(1-methylethyl)amino]methyl]-, hydrochloride (9CI), Benzyl alcohol, o-chloro-alpha-[(isopropylamino)methyl]-, hydrochloride (6CI,7CI,8CI)
InChI formel InChI=1S/C11H16ClNO.ClH/c1-8(2)13-7-11(14)9-5-3-4-6-10(9)12;/h3-6,8,11,13-14H,7H2,1-2H3;1H
Løsningstype Neat
SMIL Cl.CC(C)NCC(O)c1ccccc1Cl
IUPAC navn 1-(2-chlorophenyl)-2-(propan-2-ylamino)ethanol;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 250.16
Mængde 10 mg
Formel vægt 249.0687 g/mol