Få mere at vide
(±)-Clopidogrel-d4 Hydrogen Sulfate (2-chlorophenyl-d4), CDN
Artikelnummer. 30323501
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323501 0.005 g
30316289 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323501 Leverandør CDN Leverandørnr. CDND75010.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (RS)-Clopidogrel 2-chlorophenyl-d4 hydrogen sulfate
CAS 1219274-96-0
Molekylær formel C16 2H4 H12 Cl N O2 S . H2 O4 S
Mængde 0.005 g
Synonym Methyl 2-(2-chloro-3,4,5,6-tetradeuteriophenyl)-2-(6,7-dihydro-4 H-thieno[3,2-c]pyridin-5-yl)acetate sulfuric acid, rac-Clopidogrel-d4 hydrogen sulfate, (RS)-Methyl (2-chlorophenyl-3,4,5,6-d4)(6,7-dihydro-4 H-thieno[3,2-c]pyrid-5-yl)acetate hydrogen sulfate salt, Methyl 2-(2-Chlorophenyl-3,4,5,6-d4)-2-(6,7-dihydro-4 H-thieno[3,2-c]pyridin-5-yl)acetate hydrogen sulfate, (RS)-Clopidogrel-d4 hydrogen sulfate, (RS)-Clopidogrel-D4 hydrogen sulfate (2-chlorophenyl-D4)
InChI formel InChI=1 S/C16H16ClNO2S.H2O4S/c1-20-16(19)15(12-4-2-3-5-13(12)17)18-8-6-14-11(10-18)7-9-21-14;1-5(2,3)4/h2-5,7,9,15 H,6,8,10H2,1H3;(H2,1,2,3,4)/i2D,3 D,4 D,5 D;
SMIL [2 H]c1c([2 H])c([2 H])c(C(N2CCc3sccc3C2)C(=O)OC)c(Cl)c1[2 H].OS(=O)(=O)O
IUPAC navn methyl 2-(2-chloro-3,4,5,6-tetradeuteriophenyl)-2-(6,7-dihydro-4 H-thieno[3,2-c]pyridin-5-yl)acetate;sulfuric acid
Molekylvægt (g/mol) 423.92
Formel vægt 423.0515 g/mol
Procent renhed 98 atom % D, min 97% Chemical Purity