Få mere at vide
Clonidine Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Clonidine Hydrochloride |
| CAS | 4205-91-8 |
| Molekylær formel | C9 H9 Cl2 N3 . Cl H |
| Mængde | 50 mg |
| Synonym | 1H-Imidazol-2-amine, N-(2,6-dichlorophenyl)-4,5-dihydro-, monohydrochloride (9CI), 2-Imidazoline, 2-(2,6-dichloroanilino)-, monohydrochloride (8CI), 2-(2,6-Dichloroanilino)-2-imidazoline hydrochloride, 2-[(2,6-Dichlorophenyl)amino]-2-imidazoline hydrochloride, 2-[(2,6-Dichlorophenyl)amino]-2-imidazoline monohydrochloride, Atensina, Capresin, Catapres, Catapressan, Chlophazolin, Clofelin, Clonidine chloride, Clonidine hydrochloride, Clonidine monohydrochloride, Clopheline, DCAI, Dichloranilino imidazolin, Haemiton, Hemiton, Isoglaucon, KAPVAY, Katapresan, Neuclon, Normopresan, ST 155 |
| InChI formel | InChI=1S/C9H9Cl2N3.ClH/c10-6-2-1-3-7(11)8(6)14-9-12-4-5-13-9;/h1-3H,4-5H2,(H2,12,13,14);1H |
| SMIL | Cl.Clc1cccc(Cl)c1N=C2NCCN2 |
| IUPAC navn | N-(2,6-dichlorophenyl)imidazolidin-2-imine;hydrochloride |
| Molekylvægt (g/mol) | 266.55 |
| Formel vægt | 264.994 |