Få mere at vide
Clonidine-d4 Hydrochloride, TRC
Artikelnummer. 30368541
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30368541 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30368541 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5871321MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Clonidine-d4 Hydrochloride
CAS 67151-02-4
Molekylær formel C9 2H4 H5 Cl2 N3 . Cl H
Mængde 1 mg
Synonym Clonidine D4 hydrochloride, 1H-Imidazol-4,5-d2-2-amine, N-(2,6-dichlorophenyl)-4,5-dihydro-d2-, monohydrochloride
InChI formel InChI=1S/C9H9Cl2N3.ClH/c10-6-2-1-3-7(11)8(6)14-9-12-4-5-13-9;/h1-3H,4-5H2,(H2,12,13,14);1H/i4D2,5D2;
SMIL Cl.[2H]C1([2H])NC(=NC1([2H])[2H])Nc2c(Cl)cccc2Cl
IUPAC navn 4,4,5,5-tetradeuterio-N-(2,6-dichlorophenyl)-1H-imidazol-2-amine;hydrochloride
Molekylvægt (g/mol) 270.58
Formel vægt 269.0191