Få mere at vide
Clonidine-d4 HCl (imidazoline-4,4,5,5-d4), CDN
Artikelnummer. 30326389
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326389 0.005 g
30308969 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326389 Leverandør CDN Leverandørnr. CDND25470.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Clonidine-d4 Hydrochloride
CAS 67151-02-4
Molekylær formel C9 2H4 H5 Cl2 N3 . Cl H
Mængde 0.005 g
InChI formel InChI=1 S/C9H9Cl2N3.ClH/c10-6-2-1-3-7(11)8(6)14-9-12-4-5-13-9;/h1-3 H,4-5H2,(H2,12,13,14);1 H/i4D2,5D2;
SMIL Cl.[2 H]C1([2 H])NC(=NC1([2 H])[2 H])Nc2c(Cl)cccc2Cl
IUPAC navn 4,4,5,5-tetradeuterio-N-(2,6-dichlorophenyl)-1 H-imidazol-2-amine;hydrochloride
Molekylvægt (g/mol) 270.58
Formel vægt 269.0191 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity