Få mere at vide
Clomipramine-d6 Hydrochloride, TRC
Artikelnummer. 30360130
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30360130 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30360130 Leverandør Toronto Research Chemicals Leverandørnr. TRCC58705250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Clomipramine-d6 Hydrochloride
CAS 1189882-28-7
Renhedsgrad noter HPLC
Molekylær formel C19H17D6ClN2 • HCl
Mængde 50 mg
Synonym 3-Chloro-10,11-dihydro-N,N-(dimethyl-d6)-5H-dibenz[b,f]azepine-5-propanamine Hydrochloride, 3-Chloro-5-[3-(dimethylamino-d6)propyl]-10,11-dihydro-5H-dibenz[b,f]azepine Hydrochloride, Chlorimipramine-d6 Hydrochloride, Anafranil-d6, Clomicalm-d6
InChI formel InChI=1S/C19H23ClN2.ClH/c1-21(2)12-5-13-22-18-7-4-3-6-15(18)8-9-16-10-11-17(20)14-19(16)22;/h3-4,6-7,10-11,14H,5,8-9,12-13H2,1-2H3;1H/i1D3,2D3;
SMIL [2H]C([2H])([2H])N(CCCN1C2=CC=CC=C2CCC3=C1C=C(C=C3)Cl)C([2H])([2H])[2H].Cl
IUPAC navn 3-(2-chloro-5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-bis(trideuteriomethyl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 320.893646
Procent renhed >95