Få mere at vide
Clomipramine-d3 HCl (N-methyl-d3), CDN
Artikelnummer. 30309540
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30309540 0.005 g
30320372 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309540 Leverandør CDN Leverandørnr. CDND74660.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Clomipramine-d3 hydrochloride
CAS 1398065-86-5
Molekylær formel C19 2H3 H20 Cl N2 . Cl H
Mængde 0.005 g
Synonym Clomipramine-D3 Hydrochloride
InChI formel InChI=1 S/C19H23ClN2.ClH/c1-21(2)12-5-13-22-18-7-4-3-6-15(18)8-9-16-10-11-17(20)14-19(16)22;/h3-4,6-7,10-11,14 H,5,8-9,12-13H2,1-2H3;1 H/i1D3;
SMIL Cl.[2 H]C([2 H])([2 H])N(C)CCCN1c2ccccc2CCc3ccc(Cl)cc13
IUPAC navn 3-(2-chloro-5,6-dihydrobenzo[b][1 ]benzazepin-11-yl)-N-methyl-N-(trideuteriomethyl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 354.33
Formel vægt 353.1505 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity