Få mere at vide
Clomiphene N-Oxide, TRC
Artikelnummer. 30133882
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133882 100 mg
17940685 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133882 Leverandør Toronto Research Chemicals Leverandørnr. TRCC587040100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 97642-74-5
Molekylær formel C26 H28 Cl N O2
Molekylvægt (g/mol) 421.96
Synonym 2-[4-(2-Chloro-1,2-diphenylethenyl)phenoxy]-N,N-diethyl-ethanamine N-Oxide, Clomifenoxide
IUPAC navn 2-[4-[(E)-2-chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethylethanamine oxide
SMIL CC[N+]([O-])(CC)CCOc1ccc(cc1)\C(=C(\Cl)/c2ccccc2)\c3ccccc3
Tekniske data
Kemisk navn eller materiale Clomiphene N-Oxide
Mængde 100 mg
InChI formel InChI=1S/C26H28ClNO2/c1-3-28(29,4-2)19-20-30-24-17-15-22(16-18-24)25(21-11-7-5-8-12-21)26(27)23-13-9-6-10-14-23/h5-18H,3-4,19-20H2,1-2H3/b26-25+
Formel vægt 421.1809