Få mere at vide
Clomiphene-d5 Citrate, TRC
Artikelnummer. 30352959
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30352959 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30352959 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5870271MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Clomiphene-d5 Citrate
CAS 1217200-17-3
Renhedsgrad noter HPLC
Molekylær formel C26 D5 H23 Cl N O . C6 H8 O7
Mængde 1 mg
Synonym 2-[4-(2-Chloro-1,2-diphenylethenyl)phenoxy]-N,N-diethylethanamine 2-Hydroxy-1,2,3-propanetricarboxylate-d5, Clomid-d5, Clomphid-d5, Dyneric-d5, Fertivet-d5, Pergotime-d5, Serophene-d5, NSC 35770-d5
InChI formel InChI=1S/C26H28ClNO.C6H8O7/c1-3-28(4-2)19-20-29-24-17-15-22(16-18-24)25(21-11-7-5-8-12-21)26(27)23-13-9-6-10-14-23;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-18H,3-4,19-20H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/i6D,9D,10D,13D,14D;
SMIL [2H]c1c([2H])c([2H])c(C(=C(c2ccccc2)c3ccc(OCCN(CC)CC)cc3)Cl)c([2H])c1[2H].OC(=O)CC(O)(CC(=O)O)C(=O)O
IUPAC navn 2-[4-[2-chloro-2-(2,3,4,5,6-pentadeuteriophenyl)-1-phenyl-vinyl]phenoxy]-N,N-diethyl-ethanamine;citric acid
Molekylvægt (g/mol) 603.11
Formel vægt 602.2443
Procent renhed >95