Få mere at vide
Clomiphene citrate, Thermo Scientific™
A selective estrogen receptor modulator
Brand: Thermo Scientific Alfa Aesar J63663.09
| Mængde | 10g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsSelective estrogen receptor modulator, which acts by inhibiting the action of estrogen on the pituitary. Clomiphene Citrate is an activator of PKC and Gonad-stimulating principle.
Solubility
Slightly soluble in water, methanol, chloroform (slightly), and DMSO (10 mM). Insoluble in ether.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Kemiske identifikatorer
| 50-41-9 | |
| 598.089 | |
| PYTMYKVIJXPNBD-BTKVJIOYSA-N | |
| 6420009 | |
| CCN(CC)CCOC1=CC=C(C=C1)C(=C(C2=CC=CC=C2)Cl)C3=CC=CC=C3.C(C(=O)O)C(CC(=O)O)(C(=O)O)O |
| C32H36ClNO8 | |
| MFCD00058322 | |
| clomifene citrate, clomiphene citrate, clomid, cis-clomiphene citrate, clomiphene a citrate, zuclomid, chloramiphene, milophene, serophene, z-clomiphene citrate | |
| 2-[4-[(E)-2-chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid |
Tekniske data
| Clomiphene citrate | |
| C32H36ClNO8 | |
| 10g | |
| clomifene citrate, clomiphene citrate, clomid, cis-clomiphene citrate, clomiphene a citrate, zuclomid, chloramiphene, milophene, serophene, z-clomiphene citrate | |
| PYTMYKVIJXPNBD-BTKVJIOYSA-N | |
| 2-[4-[(E)-2-chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid | |
| 6420009 |
| 50-41-9 | |
| MFCD00058322 | |
| 14,2386 | |
| Slightly soluble in water,methanol,chloroform (slightly),and DMSO (10 mM). Insoluble in ether. | |
| CCN(CC)CCOC1=CC=C(C=C1)C(=C(C2=CC=CC=C2)Cl)C3=CC=CC=C3.C(C(=O)O)C(CC(=O)O)(C(=O)O)O | |
| 598.089 | |
| 598.08 |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.