missing translation for 'onlineSavingsMsg'
Få mere at vide

Clomiphene citrate, Thermo Scientific™

A selective estrogen receptor modulator

Brand:  Thermo Scientific Alfa Aesar J63663.04

Mængde 2g

 Se flere versioner af dette produkt

Produktkode 15455839

  • 585.83 DKK / 2g

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolitik

Beskrivelse

Beskrivelse

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Selective estrogen receptor modulator, which acts by inhibiting the action of estrogen on the pituitary. Clomiphene Citrate is an activator of PKC and Gonad-stimulating principle.

Solubility
Slightly soluble in water, methanol, chloroform (slightly), and DMSO (10 mM). Insoluble in ether.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Tekniske data

Kemiske identifikatorer

50-41-9
598.089
PYTMYKVIJXPNBD-BTKVJIOYSA-N
6420009
CCN(CC)CCOC1=CC=C(C=C1)C(=C(C2=CC=CC=C2)Cl)C3=CC=CC=C3.C(C(=O)O)C(CC(=O)O)(C(=O)O)O
C32H36ClNO8
MFCD00058322
clomifene citrate, clomiphene citrate, clomid, cis-clomiphene citrate, clomiphene a citrate, zuclomid, chloramiphene, milophene, serophene, z-clomiphene citrate
2-[4-[(E)-2-chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid

Tekniske data

Clomiphene citrate
C32H36ClNO8
2g
clomifene citrate, clomiphene citrate, clomid, cis-clomiphene citrate, clomiphene a citrate, zuclomid, chloramiphene, milophene, serophene, z-clomiphene citrate
PYTMYKVIJXPNBD-BTKVJIOYSA-N
2-[4-[(E)-2-chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
6420009
50-41-9
MFCD00058322
14,2386
Slightly soluble in water,methanol,chloroform (slightly),and DMSO (10 mM). Insoluble in ether.
CCN(CC)CCOC1=CC=C(C=C1)C(=C(C2=CC=CC=C2)Cl)C3=CC=CC=C3.C(C(=O)O)C(CC(=O)O)(C(=O)O)O
598.089
598.08
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.