missing translation for 'onlineSavingsMsg'
Få mere at vide

Clomethiazole Edisylate, Mikromol™

Artikelnummer. 17970736
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17970736 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17970736 Leverandør Mikromol Leverandørnr. MM1866.00

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Clomethiazole Edisylate
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Clomethiazole Edisylate
CAS 1867-58-9
Analyt- eller komponentnavne Clomethiazole Edisylate
Forsendelsestilstand Room Temperature
Molekylær formel 2 C6 H8 Cl N S . C2 H6 O6 S2
Certificeringer/compliance ISO 17034
Synonym 1,2-Ethanedisulfonic acid, compd. with 5-(2-chloroethyl)-4-methylthiazole (1:2), Thiazole, 5-(2-chloroethyl)-4-methyl-, 1,2-ethanedisulfonate (2:1) (8CI,9CI), Chlormethiazole edisylate, Chlormethiazole ethanedisulfonate, Clomethiazole edisylate, Clomethiazole ethane-1,2-disulfonate, Clomethiazole ethanedisulfonate, Dichlormethiazole ethanedisulfonate, Hemineurin, Hemineurine, Heminevrin, Hemithiamine, NEX 002, SCTZ, Zendra, Clomethiazole Edisylate
InChI formel InChI=1S/2C6H8ClNS.C2H6O6S2/c2*1-5-6(2-3-7)9-4-8-5;3-9(4,5)1-2-10(6,7)8/h2*4H,2-3H2,1H3;1-2H2,(H,3,4,5)(H,6,7,8)
SMIL Cc1ncsc1CCCl.Cc2ncsc2CCCl.OS(=O)(=O)CCS(=O)(=O)O
IUPAC navn 5-(2-chloroethyl)-4-methyl-1,3-thiazole;ethane-1,2-disulfonic acid
Molekylvægt (g/mol) 513.5
Mængde 100 mg
Formel vægt 511.9738 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.