Få mere at vide
Clomethiazole Edisylate, LoGiCal
Artikelnummer. 30462639
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30462639 10 mg
1 options
Artikelnummer. 30462639 Leverandør LoGiCal Leverandørnr. LGCFOR1866.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Clomethiazole Edisylate
CAS 1867-58-9
Analyt- eller komponentnavne Clomethiazole Edisylate
Forsendelsestilstand Room Temperature
Molekylær formel 2 C6 H8 Cl N S . C2 H6 O6 S2
Certificeringer/compliance ISO 9001
Synonym 1,2-Ethanedisulfonic acid, compd. with 5-(2-chloroethyl)-4-methylthiazole (1:2), Thiazole, 5-(2-chloroethyl)-4-methyl-, 1,2-ethanedisulfonate (2:1) (8CI,9CI), Chlormethiazole edisylate, Chlormethiazole ethanedisulfonate, Clomethiazole edisylate, Clomethiazole ethane-1,2-disulfonate, Clomethiazole ethanedisulfonate, Dichlormethiazole ethanedisulfonate, Hemineurin, Hemineurine, Heminevrin, Hemithiamine, NEX 002, SCTZ, Zendra, Clomethiazole Edisylate
InChI formel InChI=1S/2C6H8ClNS.C2H6O6S2/c2*1-5-6(2-3-7)9-4-8-5;3-9(4,5)1-2-10(6,7)8/h2*4H,2-3H2,1H3;1-2H2,(H,3,4,5)(H,6,7,8)
Løsningstype Neat
SMIL Cc1ncsc1CCCl.Cc2ncsc2CCCl.OS(=O)(=O)CCS(=O)(=O)O
IUPAC navn 5-(2-chloroethyl)-4-methyl-1,3-thiazole;ethane-1,2-disulfonic acid
Industry Type Clinical Testing
Molekylvægt (g/mol) 513.5
Mængde 10 mg
Formel vægt 511.9738 g/mol