missing translation for 'onlineSavingsMsg'
Få mere at vide

Clofoctol, MedChemExpress

Artikelnummer. 16428646
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16428646 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16428646 Leverandør Medchem Express Leverandørnr. HYB1150

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Clofoctol is a bacteriostatic antibiotic. Clofoctol is used in the treatment of respiratory tract and ear, nose and throat infections caused by Gram-positive bacteria. Clofoctol is only functional against Gram-positive bacteria and can penetrate into human lung tissue. Clofoctol is also an inhibitor of prostate cancer. Clofoctol has antiviral potency.

Clofoctol is a bacteriostatic antibiotic. Clofoctol is used in the treatment of respiratory tract and ear, nose and throat infections caused by Gram-positive bacteria. Clofoctol is only functional against Gram-positive bacteria and can penetrate into human lung tissue. Clofoctol is also an inhibitor of prostate cancer. Clofoctol has antiviral potency.

Tekniske data

Kemisk navn eller materiale Clofoctol
Fysisk form Solid
CAS 37693-01-9
Farve White
Mængde 100 mg
Renhedsgrad noter Research
Molekylær formel C21H26Cl2O
Til brug med (applikation) COVID-19-immunoregulation
Opløselighedsinformation DMSO : 100 mg/mL (273.72 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMIL OC1=CC=C(C(C)(C)CC(C)(C)C)C=C1CC2=CC=C(Cl)C=C2Cl
Molekylvægt (g/mol) 365.34
Formel vægt 365.34
Procent renhed 99.31%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.