Få mere at vide
Clofibric-d4 Acid, TRC
Artikelnummer. 30362389
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30362389 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30362389 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5869325MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Clofibric-d4 Acid
CAS 1184991-14-7
Molekylær formel C10 2H4 H7 Cl O3
Mængde 5 mg
Synonym 2-(4-Chloro-2,3,5,6-tetradeuteriophenoxy)-2-methylpropanoic acid, Clofibric-d4 Acid, Clofibric acid D4 (phenyl D4)
InChI formel InChI=1S/C10H11ClO3/c1-10(2,9(12)13)14-8-5-3-7(11)4-6-8/h3-6H,1-2H3,(H,12,13)/i3D,4D,5D,6D
SMIL [2H]c1c([2H])c(OC(C)(C)C(=O)O)c([2H])c([2H])c1Cl
IUPAC navn 2-(4-chloro-2,3,5,6-tetradeuteriophenoxy)-2-methylpropanoic acid
Molekylvægt (g/mol) 218.67
Formel vægt 218.0648