missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Klofibrat, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Ethyl clofibrate |
| CAS | 637-07-0 |
| Molekylær formel | C12 H15 Cl O3 |
| Mængde | 250 mg |
| Synonym | Propionic acid, 2-(p-chlorophenoxy)-2-methyl-, ethyl ester (6CI,8CI), 2-(4-Chlorophenoxy)isobutyric acid ethyl ester, 2-(p-Chlorophenoxy)-2-methylpropionic acid ethyl ester, AY 61123, Abitrate, Amotril, Anparton, Apolan, Arteriosan, Artevil, Ateculon, Ateriosan, Atheropront, Atromid S, Atromidin, Azionyl, Bioscleran, CPIB, Cartagyl, Claripex, Claripex CPIB, Clobren SF, Clofibrat, Clofibrate, Clofinit, ECPIB, EPIB, Ethyl 2-(4-chlorophenoxy)-2-methylpropionate, Ethyl 2-(4-chlorophenoxy)isobutyrate, Ethyl 2-(p-chlorophenoxy)-2-methylpropionate, Ethyl 2-(p-chlorophenoxy)isobutyrate, Ethyl clofibrate, Ethyl p-chlorophenoxyisobutyrate, Ethyl α-(4-chlorophenoxy)-α-methylpropionate, Ethyl α-(4-chlorophenoxy)isobutyrate, Ethyl α-(p-chlorophenoxy)-α-methylpropionate, Ethyl α-(p-chlorophenoxy)isobutyrate, Hyclorate, ICI 28257, Lipavil, Lipavlon, Lipomid, Liprinal, Miscleron, Misclerone, NSC 79389, Neo-Atromid, Normet, Normolipol, Recolip, Regelan, Serotinex, Sklerepmexe, Sklero-Tablinene, Sklerolip, Skleromexe, Ticlobran, Xyduril, p-Chlorophenoxyisobutyric acid ethyl ester |
| InChI formel | InChI=1S/C12H15ClO3/c1-4-15-11(14)12(2,3)16-10-7-5-9(13)6-8-10/h5-8H,4H2,1-3H3 |
| SMIL | CCOC(=O)C(C)(C)Oc1ccc(Cl)cc1 |
| IUPAC navn | ethyl 2-(4-chlorophenoxy)-2-methylpropanoat |
| Molekylvægt (g/mol) | 242.7 |
| Formel vægt | 242.071 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?