Få mere at vide
Clofibrate-d4, TRC
Artikelnummer. 30358994
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30358994 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30358994 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5869121MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Clofibrate-d4
CAS 1189654-03-2
Molekylær formel C12 2H4 H11 Cl O3
Mængde 1 mg
Synonym Propanoic acid, 2-(4-chlorophenoxy-2,3,5,6-d4)-2-methyl-, ethyl ester (ACI), Clofibric acid-d4, Ethyl 2-(4-chloro-2,3,5,6-tetradeuteriophenoxy)-2-methylpropanoate, 2-(4-Chlorophenoxy-2,3,5,6-d4)-2-methylpropanoic acid ethyl ester, Clofibrate-d4, Clofibrate-D4 (4-chlorophenyl-D4), Ethyl Clofibrate-d4 (4-chlorophenyl-d4)
InChI formel InChI=1S/C12H15ClO3/c1-4-15-11(14)12(2,3)16-10-7-5-9(13)6-8-10/h5-8H,4H2,1-3H3/i5D,6D,7D,8D
SMIL [2H]c1c([2H])c(OC(C)(C)C(=O)OCC)c([2H])c([2H])c1Cl
IUPAC navn ethyl 2-(4-chloro-2,3,5,6-tetradeuteriophenoxy)-2-methylpropanoate
Molekylvægt (g/mol) 246.72
Formel vægt 246.0961