missing translation for 'onlineSavingsMsg'
Få mere at vide

Clofibrate, 95+%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J60342.14

Mængde 25g

 Se flere versioner af dette produkt

Produktkode 15403129

  • 708.97 DKK / 25g

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolitik

Beskrivelse

Beskrivelse

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is a lipid-lowering agent used for controlling the high cholesterol and triacylglyceride level in the blood. It increases lipoprotein lipase activity to promote the conversion of VLDL to LDL, and hence reduce the level of VLDL. It can increase the level of HDL as well. Clofibrate is a ligand agonist for PPARα (peroxisome proliferator-activated receptor α), or an inducer of cytochromes P450’s CYP4A1 and CYP4A3.

Solubility
Soluble to 100 mM in DMSO.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat, bases. Store in refrigerator.
TRUSTED_SUSTAINABILITY
Tekniske data

Kemiske identifikatorer

637-07-0
242.70
KNHUKKLJHYUCFP-UHFFFAOYSA-N
2796
ethyl 2-(4-chlorophenoxy)-2-methylpropanoate
C12H15ClO3
MFCD00000615
clofibrate, ethyl clofibrate, clofibratum, clofibrato, atromid-s, miscleron, atromid, liprin, epib, angiokapsul
CHEBI:3750
CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1

Tekniske data

Clofibrate
1.14
113°C (235°F)
1.5
25g
1913459
clofibrate, ethyl clofibrate, clofibratum, clofibrato, atromid-s, miscleron, atromid, liprin, epib, angiokapsul
KNHUKKLJHYUCFP-UHFFFAOYSA-N
ethyl 2-(4-chlorophenoxy)-2-methylpropanoate
2796
242.7
637-07-0
154°C to 156°C
C12H15ClO3
MFCD00000615
UN3082
14,2377
Soluble to 100 mM in DMSO.
CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1
242.70
CHEBI:3750
≥95%
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.