Få mere at vide
Clocortolone, TRC
Artikelnummer. 30358833
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30358833 10 mg
30349721 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30358833 Leverandør Toronto Research Chemicals Leverandørnr. TRCC58380010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4828-27-7
Molekylær formel C22 H28 Cl F O4
Molekylvægt (g/mol) 410.91
Synonym Clocortolone, (6α,11β,16α)-9-Chloro-6-fluoro-11,21-dihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9-Chloro-6α-fluoro-11β,21-dihydroxy-16α-methylpregna-1,4-diene-3,20-dione, 9-Chloro-6α-fluoro-11β,21-dihydroxy-17α-methylpregna-1,4-diene-3,20-dione, 9-Chloro-6α-fluoro-16α-methyl-1,4-pregnadiene-11β,21-diol-3,20-dione, Clorcortelone
IUPAC navn (6S,8S,9R,10S,11S,13S,14S,16R,17S)-9-chloro-6-fluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-7,8,11,12,14,15,16,17-octahydro-6H-cyclopenta[a]phenanthren-3-one
SMIL C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@H]1C(=O)CO
Tekniske data
Kemisk navn eller materiale Clocortolone
Mængde 10 mg
InChI formel InChI=1S/C22H28ClFO4/c1-11-6-13-14-8-16(24)15-7-12(26)4-5-21(15,3)22(14,23)18(28)9-20(13,2)19(11)17(27)10-25/h4-5,7,11,13-14,16,18-19,25,28H,6,8-10H2,1-3H3/t11-,13+,14+,16+,18+,19-,20+,21+,22+/m1/s1
Formel vægt 410.166