Få mere at vide
Clobutinol Hydrochloride, Mikromol™
Description
Clobutinol Hydrochloride
Specifications
Specifications
| Kemisk navn eller materiale | Clobutinol Hydrochloride |
| CAS | 1215-83-4 |
| Analyt- eller komponentnavne | Clobutinol Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C14 H22 Cl N O . Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Benzeneethanol, 4-chloro-α-[2-(dimethylamino)-1-methylethyl]-α-methyl-, hydrochloride (1:1), Benzeneethanol, 4-chloro-α-[2-(dimethylamino)-1-methylethyl]-α-methyl-, hydrochloride (9CI), Phenethyl alcohol, p-chloro-α-[2-(dimethylamino)-1-methylethyl]-α-methyl-, hydrochloride (7CI,8CI), Biotertussin, Clobutinol hydrochloride, KAT 256, Silom, Silomat, Silonist |
| InChI formel | InChI=1S/C14H22ClNO.ClH/c1-11(10-16(3)4)14(2,17)9-12-5-7-13(15)8-6-12;/h5-8,11,17H,9-10H2,1-4H3;1H |
| SMIL | Cl.CC(CN(C)C)C(C)(O)Cc1ccc(Cl)cc1 |
| IUPAC navn | 1-(4-chlorophenyl)-4-(dimethylamino)-2,3-dimethylbutan-2-ol;hydrochloride |
| Show More |