missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CLK3 Recombinant Human Protein

CLK3 is a dual-specificity protein kinase.

Brand:  Invitrogen™ PV3985

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 10432523

  • 100400.00 DKK / 1mg
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CLK3 can interact with and cause the nuclear redistribution of SR proteins.

Specifications

Specifications

NP_003983, T123S, G133S
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
CLK3
Human
GST Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
86kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
CLK Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers