missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Thermo Scientific™ Human CLK2, GST Tag Recombinant Protein
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
Pakningsstørrelse:
10µg
1mg
Beskrivelse
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots; aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic.
CLK2 is a serine/threonine kinase that phosphorylates serine/arginine-rich proteins as related to controlling RNA splicing. It is also thought to link cell cycle progression, apoptosis, and telomere length regulation.
Tekniske data
Tekniske data
| Adgangsnummer | P49760 |
| Koncentration | See Label |
| Til brug med (applikation) | Kinase Assay |
| Formulering | 50mM tris with 50% glycerol, 150mM NaCl, 0.5mM EDTA, 2mM DTT, 0.02% Triton X-100 and no preservative; pH 7.5 |
| Gen-id (Entrez) | 1196 |
| Molekylvægt (g/mol) | 70.3 kDa |
| Navn | Human CLK2, GST Tag |
| pH-område | 7.5 |
| Oprensningsmetode | Purified |
| Mængde | 10 μg |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion