Få mere at vide
Clindamycin Sulfoxide (Mixture of Diastereomers), TRC
Artikelnummer. 17903095
Skift visning
Mængde:
1 mg
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17903095 1 mg -
17924995 5 mg -
17921475 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17903095 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5800101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 22431-46-5
Molekylær formel C18 H33 Cl N2 O6 S
Molekylvægt (g/mol) 440.98
Synonym L-threo-α-D-galacto-Octopyranose, 7-chloro-1,6,7,8-tetradeoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-(methylsulfinyl)-, L-threo-D-galacto-Octopyranose, 7-chloro-1,6,7,8-tetradeoxy-6-(1-methyl-4-propyl-L-2-pyrrolidinecarboxamido)-1-(methylsulfinyl)-, trans-α- (8CI), L-threo-α-D-galacto-Octopyranose, 7-chloro-1,6,7,8-tetradeoxy-6-[[(1-methyl-4-propyl-2-pyrrolidinyl)carbonyl]amino]-1-(methylsulfinyl)-, [2S-(2α,4β)]-, 7-Chloro-1,6,7,8-tetradeoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-(methylsulfinyl)-L-threo-α-D-galacto-octopyranose, 7(S)Chloro-7-deoxylincomycin sulfoxide, Clindamycin sulfoxide, U 25026A, Mixture of clindamycin sulfoxide isomers 1 & 2
IUPAC navn (2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfinyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide
SMIL CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@H](C)Cl)[C@H]2O[C@@H]([C@H](O)[C@@H](O)[C@H]2O)S(=O)C
Tekniske data
Kemisk navn eller materiale Clindamycin Sulfoxide
Renhedsgrad noter HPLC
InChI formel InChI=1S/C18H33ClN2O6S/c1-5-6-10-7-11(21(3)8-10)17(25)20-12(9(2)19)16-14(23)13(22)15(24)18(27-16)28(4)26/h9-16,18,22-24H,5-8H2,1-4H3,(H,20,25)/t9-,10+,11-,12+,13-,14+,15+,16+,18+,28?/m0/s1
Mængde 1 mg
Formel vægt 440.1748
Procent renhed >95%