Få mere at vide
Clindamycin B, TRC
Artikelnummer. 30136082
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136082 5 mg
30011321 1 mg
30009459 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136082 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5800405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 18323-43-8
Molekylær formel C17 H31 Cl N2 O5 S
Molekylvægt (g/mol) 410.96
Synonym L-threo-α-D-galacto-Octopyranoside, methyl 7-chloro-6,7,8-trideoxy-6-[[[(2S,4R)-4-ethyl-1-methyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-, L-threo-α-D-galacto-Octopyranoside, methyl 7-chloro-6,7,8-trideoxy-6-[[(4-ethyl-1-methyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-, (2S-trans)-, Methyl 7-chloro-6,7,8-trideoxy-6-[[[(2S,4R)-4-ethyl-1-methyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-L-threo-α-D-galacto-octopyranoside, Clindamycin B, Methyl 7-Chloro-6,7,8-trideoxy-6-[[[(2S,4R)-4-ethyl-1-methylpyrrolidin-2-yl]carbonyl]amino]-1-thio-L-threo-α-D-galacto-octopyranoside
IUPAC navn (2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-4-ethyl-1-methylpyrrolidine-2-carboxamide
SMIL CC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@H](C)Cl)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O
Tekniske data
Kemisk navn eller materiale Clindamycin B
InChI formel InChI=1S/C17H31ClN2O5S/c1-5-9-6-10(20(3)7-9)16(24)19-11(8(2)18)15-13(22)12(21)14(23)17(25-15)26-4/h8-15,17,21-23H,5-7H2,1-4H3,(H,19,24)/t8-,9+,10-,11+,12-,13+,14+,15+,17+/m0/s1
Mængde 5 mg
Formel vægt 410.1642