Få mere at vide
Clindamycin B 2-Phosphate, TRC
Artikelnummer. 30137579
Skift visning
Mængde:
0.5 mg
2.5 mg
5 mg
Pakningsstørrelse:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30137579 2.5 mg -
30008413 5 mg -
17903195 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137579 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5800952.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 54887-31-9
Molekylær formel C17 H32 Cl N2 O8 P S
Molekylvægt (g/mol) 490.94
Synonym (2S-trans)-Methyl 7-Chloro-6,7,8-trideoxy-6-[[(4-ethyl-1-methyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-L-threo-α-D-galacto-Octopyranoside 2-(dihydrogen phosphate);
IUPAC navn [6-[2-chloro-1-[(4-ethyl-1-methylpyrrolidine-2-carbonyl)amino]propyl]-4,5-dihydroxy-2-methylsulfanyloxan-3-yl] dihydrogen phosphate
SMIL O[C@H]1[C@@H]([C@H](NC(C2C[C@@H](CC)CN2C)=O)[C@@H](Cl)C)O[C@H](SC)C(OP(O)(O)=O)C1O
Tekniske data
Kemisk navn eller materiale Clindamycin B 2-Phosphate
InChI formel InChI=1S/C17H32ClN2O8PS/c1-5-9-6-10(20(3)7-9)16(23)19-11(8(2)18)14-12(21)13(22)15(17(27-14)30-4)28-29(24,25)26/h8-15,17,21-22H,5-7H2,1-4H3,(H,19,23)(H2,24,25,26)
Mængde 2.5 mg
Formel vægt 490.1306