Få mere at vide
Clindamycin-13C,D3, TRC
Artikelnummer. 30358503
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30358503 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30358503 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5799925MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Clindamycin-13C,D3
CAS 2140264-63-5
Renhedsgrad noter HPLC
Molekylær formel 13C C17 D3 H30 Cl N2 O5 S
Mængde 5 mg
Synonym Methyl 7-Chloro-6,7,8-trideoxy-6-[[[(2S,4R)-1-methyl-13C,D3-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-L-threo-α-D-galacto-octopyranoside, trans-α-Methyl 7-Chloro-6,7,8-trideoxy-6-(1-methyl-13C,D3-4-propyl-L-2-pyrrolidinecarboxamido)-1-thio-L-threo-D-galacto-octopyranoside, (2S-trans)-Methyl 7-Chloro-6,7,8-trideoxy-6-[[(1-methyl-13C,D3-4-propyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-L-threo-α-D-galacto-octopyranoside, 7(S)-Chloro-7-deoxylincomycin-13C,D3, 7-CDL-13C,D3, 7-Chloro-7-deoxylincomycin-13C,D3, 7-Chlorolincomycin-13C,D3, 7-Deoxy-7(S)-chlorolincomycin-13C,D3, Antirobe-13C,D3, Chlolincocin-13C,D3, Cleocin-13C,D3, Clincin-13C,D3, ClindaDerm-13C,D3, Clindam-13C,D3, Clindamycin-13C,D3, Clinimycin-13C,D3, Dalacin C-13C,D3, Dalacin V-13C,D3, Dalacine-13C,D3, Klimicin-13C,D3, Klindan 3
InChI formel InChI=1S/C18H33ClN2O5S/c1-5-6-10-7-11(21(3)8-10)17(25)20-12(9(2)19)16-14(23)13(22)15(24)18(26-16)27-4/h9-16,18,22-24H,5-8H2,1-4H3,(H,20,25)/t9-,10+,11-,12+,13-,14+,15+,16+,18+/m0/s1/i3+1D3
SMIL [2H][13C]([2H])([2H])N1C[C@H](CCC)C[C@H]1C(=O)N[C@H]([C@H](C)Cl)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O
IUPAC navn (2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyl-tetrahydropyran-2-yl]propyl]-4-propyl-1-(trideuterio(1^{13}C)methyl)pyrrolidine-2-carboxamide
Molekylvægt (g/mol) 428.99
Formel vægt 428.2021
Procent renhed >95