missing translation for 'onlineSavingsMsg'
Få mere at vide

Clindamycin-13C,D3 Hydrochloride, TRC

Artikelnummer. 30362411
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.5 mg
5 mg
Pakningsstørrelse:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30362411 5 mg -
30351168 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30362411 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5799945MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 2140264-64-6
Molekylær formel C1713CH30D3ClN2O5S • HCl
Molekylvægt (g/mol) 465.45
Synonym Methyl 7-Chloro-6,7,8-trideoxy-6-[[[(2S,4R)-1-methyl-13C,D3-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-L-threo-α-D-galacto-octopyranoside Hydrochloride, trans-α-Methyl 7-Chloro-6,7,8-trideoxy-6-(1-methyl-13C,D3-4-propyl-L-2-pyrrolidinecarboxamido)-1-thio-L-threo-D-galacto-octopyranoside Hydrochloride, (2S-trans)-Methyl 7-Chloro-6,7,8-trideoxy-6-[[(1-methyl-13C,D3-4-propyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-L-threo-α-D-galacto-octopyranoside Hydrochloride, 7(S)-Chloro-7-deoxylincomycin-13C,D3 Hydrochloride, 7-CDL-13C,D3 Hydrochloride, 7-Chloro-7-deoxylincomycin-13C,D3 Hydrochloride, 7-Chlorolincomycin-13C,D3 Hydrochloride, 7-Deoxy-7(S)-chlorolincomycin-13C,D3 Hydrochloride, Antirobe-13C,D3 Hydrochloride, Chlolincocin-13C,D3 Hydrochloride, Cleocin-13C,D3 Hydrochloride, Clincin-13C,D3 Hydrochloride, ClindaDerm-13C,D3 Hydrochloride, Clindam-13C,D3 Hydrochloride, Clindamycin-13C,D3 Hydrochloride, Clinimycin-13C,D3 Hydrochloride, Dalacin C-13C,D3 Hydrochloride, Dalacin V-13C,D3 Hydrochloride, Dalacine-13C,D3 Hydrochloride, Klimicin-13C,D3 Hydrochloride, Klindan 300-13C,D3 Hydrochloride, Sobelin-13C,D3 Hydrochloride, U 21251-13C,D3 Hydrochloride
IUPAC navn (2S,4R)-N-((1S,2S)-2-chloro-1-((2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(methylthio)tetrahydro-2H-pyran-2-yl)propyl)-1-(methyl-13C-d3)-4-propylpyrrolidine-2-carboxamide hydrogen chloride
SMIL CCC[C@@H]1C[C@@H](C(N[C@@H]([C@@H]2[C@H](O)[C@H](O)[C@@H](O)[C@@H](SC)O2)[C@@H](Cl)C)=O)N([13C]([2H])([2H])[2H])C1.Cl

Tekniske data

Kemisk navn eller materiale Clindamycin-13C,D3 Hydrochloride
Mængde 5 mg
InChI formel InChI=1S/C18H33ClN2O5S.ClH/c1-5-6-10-7-11(21(3)8-10)17(25)20-12(9(2)19)16-14(23)13(22)15(24)18(26-16)27-4;/h9-16,18,22-24H,5-8H2,1-4H3,(H,20,25);1H/t9-,10+,11-,12+,13-,14+,15+,16+,18+;/m0./s1/i3+1D3;
Formel vægt 464.19
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.