Learn More
Nickel Electro™ Clifton™ Stainless Steel Flat Lid
Maintain a stable bath temperature and reduce evaporation with a stainless steel lid. Nickel Electro™ Clifton™ Stainless Steel Flat Lid minimises heat loss and reduces evaporation.
Brand: Nickel Electro™ LD-90
Additional Details : Weight : 0.50000kg
Description
- Stainless steel
- Insulated
- Minimises heat loss
- Reduces evaporation
- Assists in maintaining bath temperature
Specifications
90L Baths | |
Stainless Steel |