missing translation for 'onlineSavingsMsg'
Få mere at vide

Clevidipine, TRC

Artikelnummer. 30138752
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138752 25 mg
30136564 100 mg
17912025 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30138752 Leverandør Toronto Research Chemicals Leverandørnr. TRCC57370025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 167221-71-8
Molekylær formel C21H23Cl2NO6
Molekylvægt (g/mol) 456.32
Synonym 4-(2,3-Dichlorophenyl)-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid 3-Methyl 5-[(1-Oxobutoxy)methyl] Ester, Clevelox, Cleviprex, H 324/38, rac-Clevidipine;
IUPAC navn 5-O-(butanoyloxymethyl) 3-O-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMIL CCCC(=O)OCOC(=O)C1=C(C)NC(=C(C1C2=C(C(=CC=C2)Cl)Cl)C(=O)OC)C

Specifications

Kemisk navn eller materiale Clevidipine
Mængde 25 mg
InChI formel InChI=1S/C21H23Cl2NO6/c1-5-7-15(25)29-10-30-21(27)17-12(3)24-11(2)16(20(26)28-4)18(17)13-8-6-9-14(22)19(13)23/h6,8-9,18,24H,5,7,10H2,1-4H3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.