Få mere at vide
Cletoquine-D4 Oxalate, TRC
Artikelnummer. 30363455
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30363455 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30363455 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5735021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cletoquine-d4 Oxalate
CAS 1216461-57-2
Molekylær formel C18H20D4ClN3O5
Mængde 1 mg
Synonym 2-[[4-[(7-Chloroquinolin-4-yl)amino]-1,1,2,2-tetradeuteriopentyl]amino]ethanol oxalate, 2-[[4-[(7-Chloro-4-quinolinyl)amino]pentyl-1,1,2,2-d4]amino]ethanol ethanedioate (1:1), Cletoquine-d4 oxalate, Desethylhydroxychloroquine-d4 oxalate, Desethylhydroxychloroquine-D4 (1,4-diaminopentyl-1,1,2,2-D4) oxalate
InChI formel InChI=1S/C16H22ClN3O.C2H2O4/c1-12(3-2-7-18-9-10-21)20-15-6-8-19-16-11-13(17)4-5-14(15)16;3-1(4)2(5)6/h4-6,8,11-12,18,21H,2-3,7,9-10H2,1H3,(H,19,20);(H,3,4)(H,5,6)/i2D2,7D2;
SMIL [2H]C([2H])(CC(C)Nc1ccnc2cc(Cl)ccc12)C([2H])([2H])NCCO.OC(=O)C(=O)O
IUPAC navn 2-[[4-[(7-chloroquinolin-4-yl)amino]-1,1,2,2-tetradeuteriopentyl]amino]ethanol;oxalic acid
Molekylvægt (g/mol) 401.88