Få mere at vide
Clethodim Sulfone, TRC
Artikelnummer. 17904915
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17904915 10 mg
17960995 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17904915 Leverandør Toronto Research Chemicals Leverandørnr. TRCC57326010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 111031-17-5
Molekylær formel C17 H26 Cl N O5 S
Molekylvægt (g/mol) 391.91
Synonym 2-Cyclohexen-1-one, 2-[1-[[(3-chloro-2-propenyl)oxy]imino]propyl]-5-[2-(ethylsulfonyl)propyl]-3-hydroxy- (9CI), 2-[1-[[(3-Chloro-2-propen-1-yl)oxy]imino]propyl]-5-[2-(ethylsulfonyl)propyl]-3-hydroxy-2-cyclohexen-1-one, Clethodim sulfone
IUPAC navn 2-[(E)-N-[(E)-3-chloroprop-2-enoxy]-C-ethylcarbonimidoyl]-5-(2-ethylsulfonylpropyl)-3-hydroxycyclohex-2-en-1-one
SMIL CC\C(=N/OC\C=C\Cl)\C1=C(O)CC(CC(C)S(=O)(=O)CC)CC1=O
Tekniske data
Kemisk navn eller materiale Clethodim Sulfone
Analyt- eller komponentnavne Clethodim Sulfone
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C17H26ClNO5S/c1-4-14(19-24-8-6-7-18)17-15(20)10-13(11-16(17)21)9-12(3)25(22,23)5-2/h6-7,12-13,20H,4-5,8-11H2,1-3H3/b7-6+,19-14+
Løsningstype Neat
Industry Type Forensics
Mængde 10 mg
Formel vægt 391.122