Få mere at vide
Clencyclohexerol-d10, TRC
Artikelnummer. 30366683
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30366683 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366683 Leverandør Toronto Research Chemicals Leverandørnr. TRCC57250210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Clencyclohexerol-d10
CAS 1346599-61-8
Renhedsgrad noter HPLC
Molekylær formel C14H10D10Cl2N2O2
Mængde 10 mg
Synonym 4-Amino-3,5-dichloro-α-[[(4-hydroxycyclohexyl-d10)amino]methyl]benzenemethanol
InChI formel InChI=1S/C14H20Cl2N2O2/c15-11-5-8(6-12(16)14(11)17)13(20)7-18-9-1-3-10(19)4-2-9/h5-6,9-10,13,18-20H,1-4,7,17H2/i1D2,2D2,3D2,4D2,9D,10D
SMIL OC1([2H])C([2H])([2H])C([2H])([2H])C(NCC(O)C2=CC(Cl)=C(N)C(Cl)=C2)([2H])C([2H])([2H])C1([2H])[2H]
IUPAC navn 4-[[2-(4-amino-3,5-dichlorophenyl)-2-hydroxyethyl]amino]-1,2,2,3,3,4,5,5,6,6-decadeuteriocyclohexan-1-ol
Molekylvægt (g/mol) 329.29
Formel vægt 328.153
Procent renhed >95