missing translation for 'onlineSavingsMsg'
Få mere at vide

Clemizole hydrochloride, MedChemExpress

Artikelnummer. 16493947
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16493947 50 mg
16448028 10 mg
16440488 5 mg
16417806 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16493947 Leverandør Medchem Express Leverandørnr. HY30234A

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Clemizole hydrochloride is an H1 histamine receptor antagonist, is found to substantially inhibit HCV replication. Clemizole hydrochloride is an inhibitor of TRPC5 channel. The IC50 of Clemizole hydrochloride for RNA binding by NS4B is 24 nM, whereas its EC50 for viral replication is 8 μM.

Clemizole hydrochloride is an H1 histamine receptor antagonist, is found to substantially inhibit HCV replication. Clemizole hydrochloride is an inhibitor of TRPC5 channel. The IC50 of Clemizole hydrochloride for RNA binding by NS4B is 24 nM, whereas its EC50 for viral replication is 8 μM.

Tekniske data

Kemisk navn eller materiale Clemizole hydrochloride
CAS 1163-36-6
Farve White
Renhedsgrad noter Research
Molekylær formel C19H21Cl2N3
Mængde 50 mg
Opløselighedsinformation DMSO : 33.33 mg/mL (92.00 mM; Need ultrasonic) ∣H2O : 20 mg/mL (55.20 mM; Need ultrasonic)
SMIL ClC1=CC=C(C=C1)CN2C(CN3CCCC3)=NC4=CC=CC=C24.[H]Cl
Molekylvægt (g/mol) 362.3
Formel vægt 362.3
Procent renhed 99.24%
Grad Research
Fysisk form Powder
Til brug med (applikation) COVID-19-anti-virus
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.