Få mere at vide
Clemizole Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Clemizole Hydrochloride |
| CAS | 1163-36-6 |
| Analyt- eller komponentnavne | Clemizole Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C19 H20 Cl N3 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1H-Benzimidazole, 1-[(4-chlorophenyl)methyl]-2-(1-pyrrolidinylmethyl)-, hydrochloride (1:1), 1H-Benzimidazole, 1-[(4-chlorophenyl)methyl]-2-(1-pyrrolidinylmethyl)-, monohydrochloride (9CI), Benzimidazole, 1-(p-chlorobenzyl)-2-(1-pyrrolidinylmethyl)-, monohydrochloride (8CI), 1-(4-Chlorobenzyl)-2-(pyrrolidinylmethyl)-1,3-benzimidazole hydrochloride, 1-p-Chlorobenzyl-2-(1-pyrrolidinylmethyl)benzimidazole hydrochloride, 1-p-Chlorobenzyl-2-pyrrolidylmethyl benzimidazole hydrochloride, AL 20, Allercur, Clemizole hydrochloride, Histacuran, Klemidox, Reactrol, Clemizole Hydrochloride |
| InChI formel | InChI=1S/C19H20ClN3.ClH/c20-16-9-7-15(8-10-16)13-23-18-6-2-1-5-17(18)21-19(23)14-22-11-3-4-12-22;/h1-2,5-10H,3-4,11-14H2;1H |
| Løsningstype | Neat |
| SMIL | Cl.Clc1ccc(Cn2c(CN3CCCC3)nc4ccccc24)cc1 |
| Vis mere |