missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CLEC3B/Tetranectin Antibody (011), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90684FR
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CLEC3B/Tetranectin Monoclonal specifically detects CLEC3B/Tetranectin in Mouse samples. It is validated for ELISA.
Tekniske data
| CLEC3B/Tetranectin | |
| Monoclonal | |
| DyLight 680 | |
| C-type lectin domain family 3 member B, C-type lectin domain family 3, member B, DKFZp686H17246, Plasminogen kringle 4-binding protein, tetranectin (plasminogen binding protein), TNAtetranectin (plasminogen-binding protein), TNtetranectin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CLEC3B/Tetranectin (P43025; Met1-Val202). | |
| 0.1 mL | |
| Cancer | |
| 7123 | |
| Mouse | |
| Purified |
| ELISA | |
| 11 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Mouse CLEC10A Mouse CLEC14A | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion