Få mere at vide
Diversey™ Clearklens Bi-Spore Disinfectant
Artikelnummer. 15340308
Volumen (metrisk):
20 l
Pakningsstørrelse:
20 L
Artikelnummer. 15340308
Brand: Diversey™ 7518163
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Disinfects hard surfaces with a sporicidal solution. Diversey Clearklens Bi-Sore Disinfectant is a chlorine dioxide generating system. It is effective against all types of micro-organisms.
  • Effective on hard surfaces against all types of micro-organisms, including bacteria, yeasts, fungi, spores, and viruses
  • Aseptically filled, two-part system for the creation of chlorine dioxide presented as an activator and base concentrate solutions
  • Double bagged to avoid environmental contamination
Tekniske data
Sterilitet Sterile
Filtreret Filtered
Volumen (metrisk) 20 l