Få mere at vide
Berlin Packaging™ Clear Laboratory Glassware Pot
Artikelnummer. 30408780
Skift visning
Kapacitet (metrisk):
100 mL
250 mL
500 mL
1000 mL
Mængde:
18 Set
20 Set
48 Set
63 Set
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Kapacitet (metrisk) Mængde
30408780 1000 mL 18 Set
30408786 100 mL 63 Set
30408783 250 mL 48 Set
30408784 500 mL 20 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 30408780 Leverandør Berlin Packaging™ Leverandørnr. 1000732
  • ONLINE EXCLUSIVE PRICE
    23585.31 DKK
    18929.20 DKK / Pakke med 720
    Spare 4656.11 DKK
    19% Slukket
Forventet forsendelse: 07-09-2026
Læg i indkøbskurv
Læg i indkøbskurv
Use the Berlin Packaging™ Clear Laboratory Glassware Pot as a fundamental piece of laboratory glassware. The autoclavable clear glass is breakproof and chemically and thermally shockproof.
  • Clear borosilicate glass
  • Break-resistant
  • Chemically and thermally shockproof;
  • In compliance with EP, USP and JP guidelines
  • Autoclavable
Tekniske data
Certificeringer/compliance DIN 68
Mængde 18 Set
Kapacitet (metrisk) 1000 mL
Materiale Glass
Produkttype Pot