Beskrivelse
- Tackle a range of tasks with our Tork Cleaning Cloth - it's suitable with most solvents, disinfectants and detergents.
- Replace unsightly rags and rental towels with these durable cloths for repeated use.
- Swiftly remove oil, grease, lubricants, and dirt, thanks to their high absorbency.
- Effortlessly complete lighter cleaning tasks with its soft and flexible feel.
Tekniske data
Tekniske data
| Produkttype | Cleaning Cloth |
| Længde (engelsk) | 166.23 yd. |
| Længde (metrisk) | 152 m |
| Bredde (engelsk) | 12.4 in. |
| Bredde (metrisk) | 31.5 cm |
| Autoklaverbar | Not Autoclavable |
| Certificeringer/compliance | FSC MIX Credit, ISO 9001, ISO 14001 |
| Farve | Blå |
| Engangs | Limited-use |
| Emballagemængde | 1 Roll |
| Vis mere |