Få mere at vide
Emma Safety Footwear Clay (Ruffneck) Safety Shoes
Størrelse:
39
40
41
42
43
45
46
47
48
49
Pakningsstørrelse:
1 par
Beskrivelse
- S3-Safety shoe with sole plate for wet work
- Nubuck / Oil Nubuck construction
- GripForce™ Easy Twist Outsole
- 3M™ Reflective Striping
- ESD certified
- Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Tekniske data
Tekniske data
| Certificeringer/compliance | EN ISO 20345:2011, EN 12568:2010 |
| Lukketype | Laces |
| Farve | Black |
| Design | Clay (Ruffneck) |
| Liner materiale | Hydro-Tec™ Sanitized Silver |
| Materiale | Nubuck, Oiled Nubuck |
| Størrelse | 42 |
| Tå materiale | Steel |