Få mere at vide
Clavulanic Acid Methyl-d3 Ester, TRC
Artikelnummer. 30349254
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30349254 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30349254 Leverandør Toronto Research Chemicals Leverandørnr. TRCC56377250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Clavulanic Acid Methyl-d3 Ester
CAS 1821757-56-5
Renhedsgrad noter HPLC
Molekylær formel C9H8D3NO5
Mængde 50 mg
Synonym (2R,3Z,5R)-3-(2-Hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Methyl-d3 Ester, [2R-(2α,3Z,5α)]- 3-(2-Hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Methyl-d3 Ester, Methyl-d3 Clavulanate
InChI formel InChI=1S/C9H11NO5/c1-14-9(13)8-5(2-3-11)15-7-4-6(12)10(7)8/h2,7-8,11H,3-4H2,1H3/b5-2-/t7-,8-/m1/s1/i1D3
SMIL O=C1C[C@@]2(N1[C@](/C(O2)=C/CO)(C(OC([2H])([2H])[2H])=O)[H])[H]
IUPAC navn methyl-d3 (2R,5R,Z)-3-(2-hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molekylvægt (g/mol) 216.21
Formel vægt 216.08
Procent renhed >95