Få mere at vide
Clavulanate Lithium, TRC
Artikelnummer. 30137543
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137543 1 g
17914435 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137543 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5637401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61177-44-4
Molekylær formel C8 H8 N O5 . Li
Molekylvægt (g/mol) 205.09
Synonym (2R,3Z,5R)-3-(2-hydroxyethylidene)-7-oxo-4-Oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic acid lithium salt (1:1), (2R,3Z,5R)-3-(2-hydroxyethylidene)-7-oxo-4-Oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic acid lithium salt (1:1), 4-Oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3-(2-hydroxyethylidene)-7-oxo-, monolithium salt, (2R,3Z,5R)- (9CI), 4-Oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3-(2-hydroxyethylidene)-7-oxo-, monolithium salt, [2R-(2α,3Z,5α)]-, Lithium clavulanate Clavulanate Lithium
IUPAC navn lithium;(2R,3Z,5R)-3-(2-hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMIL [Li+].OC\C=C\1/O[C@@H]2CC(=O)N2[C@H]1C(=O)[O-]
Tekniske data
Kemisk navn eller materiale Lithium clavulanate
Mængde 1 g
InChI formel InChI=1S/C8H9NO5.Li/c10-2-1-4-7(8(12)13)9-5(11)3-6(9)14-4;/h1,6-7,10H,2-3H2,(H,12,13);/q;+1/p-1/b4-1-;/t6-,7-;/m1./s1
Formel vægt 205.0563