Få mere at vide
Novus Biologicals™ Claudin-9 Overexpression Lysate
Artikelnummer. 18277841 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mg
Artikelnummer. Mængde
18277841 0.1 mg
1 options
Artikelnummer. 18277841 Leverandør Novus Biologicals™ Leverandørnr. NBL109251
 Begrænset vare
Forventet forsendelse: 15-10-2026
Læg i indkøbskurv
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
The Claudin-9 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Cellelinje The lysate was created in HEK293T cells, using plasmid ID RC2871 and based on accession number NM_2982. The protein contains a C-terminal DDK tag.
Til brug med (applikation) Western Blot
Mængde 0.1 mg
Opbevaringsbuffer RIPA buffer
Renhed Protein
Testspecificitet Homo sapiens claudin 9 (CLDN9), mRNA.
Indhold og opbevaring -80°C. Avoid freeze-thaw cycles.
Proteintilstand Native
For Research Use Only