Få mere at vide
Prestige Medical™ Classic Portable Autoclave Media
Artikelnummer. 12745355
Kapacitet (metrisk):
9 og 12 l
Pakningsstørrelse:
Hver
Artikelnummer. 12745355
Brand: Prestige Medical™ 210047UK
The availability of a wide range of model sizes, features and internal furniture options make the Prestige Medical Classic a popular choice where a cost effective, portable autoclave is needed. This 9 liter Classic Media autoclave sterilises at 121°C and has a cycle time of approximately 70 minutes. Fully automatic, simply press the button to start. Lightweight and portable, ideal for laboratory use - the appliance for science.
  • High quality construction
  • Fully automatic electronic time and temperature control
  • Steam release valve automatically opens to release excess pressure
  • Automatic fault warning light and cut-out if unit boils dry or sterilisation is interrupted
  • Dual interlock handles prevent the lid being opened before the pressure and temperature inside have fallen to safe levels
  • Complete with instrument basket
  • Models 2015 onwards available with optional printer for easy data recording
  • CE marked
  • Comprehensive one year warranty (UK)
  • Interlock safety mechanism, eliminates the risk of super-heated vessels exploding
  • Fully automatic, 'Set and forget' - load your flask into the basket, set the programme with a one touch button, relax and forget until the cycle has ended
  • Reliable, special in-built software ensures exact temperatures are reached for the correct duration
  • 9L and 12L models
  • Fits neatly onto the smallest of workbenches For the sterilisation of media, liquids and contaminated waste.
Tekniske data
Kapacitet (metrisk) 9 og 12 l