Learn More
Zwiebel™ Class M2 Chrome Weight Set
Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M2 Weights are ideal for the verification or calibration of class M3 weights.
Brand:  Zwiebel™ PBI23705
 View more versions of this product
Product Code. 11724714
  • 2355.00 DKK / Each
Only null left
Estimated Shipment: 07-08-2025
Description
  • Class M2 stainless steel calibration weight
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
1 to 1000 g
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Zwiebel™ Class M2 Chrome Weight Set > Mass: 1 to 1000g