Learn More
Zwiebel™ Plastic case for 20kg Precision Weight
Empty plastic carry case for 20 kg weight, rectangular shape.
Brand: Zwiebel™ PBI23836
This item is not returnable.
View return policy
Description
- Holds one M1 weight
Specifications
Weight | |
M1 | |
20 kg |
Product Content Correction
Product Title
Zwiebel™ Plastic case for 20kg Precision Weight > 20 kg weight
Spot an opportunity for improvement?Share a Content Correction